siRNA Details
| Gene Symbol: Olfr555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147103.2 | NP_667314.2 | 1 | 635 | ||
| GenBank mRNA | BC106981.1 | - | 1 | 635 | 40043127 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr555| Gene Symbol: Olfr555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147103.2 | NP_667314.2 | 1 | 635 | ||
| GenBank mRNA | BC106981.1 | - | 1 | 635 | 40043127 | |