siRNA Details
| Gene Symbol: Nr5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139051.3 | NP_620639.1 | 7 | 1409 | ||
| GenBank mRNA | AB000490.1 | - | 7 | 1453 | ||
| AF511594.1 | - | 7 | 1239 | |||
| AK134378.1 | - | 7 | 1410 | |||
| AK144376.1 | - | 7 | 1370 | |||
| AK156911.1 | - | 7 | 1377 | |||
| BC110476.1 | - | 7 | 1352 | 40047261 | ||
| BC110477.1 | - | 7 | 1352 | 40047262 | ||

Nr5a1
Genomic Map