siRNA Details
| Gene Symbol: Ly6g5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148939.2 | NP_683741.1 | 2 | 169 | ||
| GenBank mRNA | AJ315553.1 | - | 2 | 169 | ||
| AJ315554.1 | - | 1 | 265 | |||
| AK151902.1 | - | 1 | 206 | |||
| BC131964.1 | - | 2 | 197 | 40130241 | ||
| BC131966.1 | - | 2 | 197 | 40130243 | ||
| BC145150.1 | - | 1 | 293 | |||
| HE864438.1 | - | 2 | 185 | |||
| HE864439.1 | - | 1 | 281 | |||

Ly6g5b
Genomic Map