siRNA Details
| Gene Symbol: A930011O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK044422.1 | - | LIT3369 RNCR3 |
2 | 56 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930011O12Rik| Gene Symbol: A930011O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK044422.1 | - | LIT3369 RNCR3 |
2 | 56 | |