siRNA Details
Gene Symbol: Robo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175549.4 | NP_780758.3 | 5 | 684 | ||
GenBank mRNA | AK035110.1 | - | 5 | 1558 | ||
AK085461.1 | - | 5 | 1309 | |||
BC055333.1 | - | 5 | 992 | 6409283 | ||
DQ533875.1 | - | 6 | 964 | |||
DQ533876.1 | - | 5 | 684 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK164890.1 | - | 1733 | Chr. - - [-] on Build |
