siRNA Details
| Gene Symbol: Cops7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172974.2 | NP_766562.1 | 5 | 552 | ||
| GenBank mRNA | AF071317.1 | - | 5 | 530 | ||
| AK053615.1 | - | 4 | 422 | |||
| AK081309.1 | - | 5 | 552 | |||
| AK168740.1 | - | 5 | 511 | |||
| AK170140.1 | - | 5 | 511 | |||
| AK182444.1 | - | 2 | 174 | |||
| AK205287.1 | - | 2 | 174 | |||
| AK207190.1 | - | 1 | 85 | |||
| AK208348.1 | - | 2 | 174 | |||
| AK214314.1 | - | 2 | 174 | |||
| AK214430.1 | - | 2 | 174 | |||
| BC012659.1 | - | 5 | 516 | 4017012 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108807, XM_006245415, XM_006245414.

Cops7b
Genomic Map