siRNA Details
| Gene Symbol: Slc28a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172980.2 | NP_766568.1 | 6, 7 | 1790 | ||
| GenBank mRNA | AK080957.1 | - | 6, 7 | 1791 | ||
| AK152776.1 | - | 5, 6 | 725 | |||
| BC055376.1 | - | 5, 6 | 684 | 30077988 | ||

Slc28a2
Genomic Map