siRNA Details
| Gene Symbol: Vsnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012038.4 | NP_036168.1 | 4 | 747 | ||
| GenBank mRNA | AJ489331.1 | - | 4 | 711 | ||
| AK005091.2 | - | 3 | 309 | |||
| AK144454.1 | - | 4 | 677 | |||
| AK160224.1 | - | 4 | 679 | |||
| AY101375.1 | - | 3 | 449 | |||
| BC046226.1 | - | 4 | 652 | 5692284 | ||
| D21165.1 | - | 3 | 449 | |||
| EU373446.1 | - | 3 | 449 | |||

Vsnl1
Genomic Map