siRNA Details
| Gene Symbol: Gsg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101488.1 | NP_001094958.1 | 1, 2 | 560 | ||
| GenBank mRNA | AK082731.1 | - | 1, 2 | 562 | ||
| BB651613.1 | - | 1, 2 | 555 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsg1l| Gene Symbol: Gsg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101488.1 | NP_001094958.1 | 1, 2 | 560 | ||
| GenBank mRNA | AK082731.1 | - | 1, 2 | 562 | ||
| BB651613.1 | - | 1, 2 | 555 | |||