siRNA Details
| Gene Symbol: Pcnxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175561.4 | NP_780770.2 | 32 | 5850 | ||
| GenBank mRNA | AK030215.1 | - | 7 | 1394 | ||
| AK087907.1 | - | 17 | 2766 | |||
| AK220342.1 | - | 24 | 3412 | |||
| BC068235.1 | - | 21 | 3917 | |||

Pcnxl2
Genomic Map