siRNA Details
| Gene Symbol: Pcnxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175561.4 | NP_780770.2 | 33 | 6321 | ||
| GenBank mRNA | AK030215.1 | - | 8 | 1865 | ||
| AK087907.1 | - | 18 | 3237 | |||
| AK220342.1 | - | 25 | 3883 | |||
| BC068235.1 | - | 22 | 4388 | |||

Pcnxl2
Genomic Map