siRNA Details
| Gene Symbol: Pcnxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175561.4 | NP_780770.2 | 31 | 5760 | ||
| GenBank mRNA | AK030215.1 | - | 6 | 1304 | ||
| AK087907.1 | - | 16 | 2676 | |||
| AK220342.1 | - | 23 | 3322 | |||
| BC068235.1 | - | 20 | 3827 | |||

Pcnxl2
Genomic Map