siRNA Details
| Gene Symbol: Sgsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018822.3 | NP_061292.2 | 8 | 1336 | ||
| GenBank mRNA | AF153827.1 | - | 8 | 1310 | ||
| AF304053.1 | - | 8 | 1310 | |||
| AF304054.1 | - | 8 | 1310 | |||
| AK034220.1 | - | 8 | 1140 | |||
| AK052266.1 | - | 8 | 1329 | |||
| AK076267.1 | - | 8 | 1366 | |||
| AK128926.1 | - | 8 | 1244 | |||
| AK141266.1 | - | 8 | 1284 | |||
| BC130268.1 | - | 8 | 1333 | 40143074 | ||

Sgsh
Genomic Map