siRNA Details
| Gene Symbol: Bcap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012060.4 | NP_036190.2 | 4 | 322 | ||
| GenBank mRNA | AK008043.1 | - | 4 | 322 | ||
| AK011500.1 | - | 4 | 323 | |||
| AK151641.1 | - | 4 | 323 | |||
| BC002106.1 | - | 4 | 290 | 3486784 | ||
| X81816.1 | - | 3 | 222 | |||

Bcap31
Genomic Map