siRNA Details
| Gene Symbol: Zfp275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160229.1 | NP_001153701.1 | 3, 4 | 332 | ||
| NM_031494.2 | NP_113682.2 | 4, 5 | 425 | |||
| GenBank mRNA | AB221659.1 | - | 1, 2 | 140 | ||
| AK142957.1 | - | 3, 4 | 332 | |||
| BC138002.1 | - | 4, 5 | 330 | 8861021 | ||
| BC138003.1 | - | 4, 5 | 330 | 8861022 | ||
| CJ130997.1 | - | 4, 5 | 427 | |||

Zfp275
Genomic Map