siRNA Details
| Gene Symbol: Cd200r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206535.1 | NP_996258.1 | 5, 6 | 949 | ||
| GenBank mRNA | AY230198.1 | - | 5, 6 | 949 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd200r2| Gene Symbol: Cd200r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206535.1 | NP_996258.1 | 5, 6 | 949 | ||
| GenBank mRNA | AY230198.1 | - | 5, 6 | 949 | ||