siRNA Details
| Gene Symbol: Rbbp8nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173031.3 | NP_766619.2 | 2 | 189 | ||
| GenBank mRNA | AK028610.1 | - | 2 | 191 | ||
| BC066135.1 | - | 2 | 145 | |||
| BC150940.1 | - | 2 | 144 | |||
| BC172093.1 | - | 2 | 144 | |||
| BY714078.1 | - | 2 | 191 | |||

Rbbp8nl
Genomic Map