siRNA Details
| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143671.1 | NP_001137143.1 | 8 | 873 | ||
| NM_001143672.1 | NP_001137144.1 | 7 | 810 | |||
| GenBank mRNA | AK131817.1 | - | 8 | 874 | ||
| BC039763.1 | - | 6 | 637 | |||
| BC042836.1 | - | 6 | 626 | |||
| BC089620.1 | - | 7 | 785 | |||

Tmem255b
Genomic Map