siRNA Details
Gene Symbol: Dok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013739.2 | NP_038767.1 | 1, 2 | 96 | ||
GenBank mRNA | AF179242.1 | - | 1, 2 | 96 | ||
AF237580.1 | - | 1, 2 | 92 | |||
AK137520.1 | - | 1, 2 | 83 | |||
BC116922.1 | - | 1, 2 | 95 | 40126241 | ||
BC120535.1 | - | 1, 2 | 95 | 40129458 |
