siRNA Details
| Gene Symbol: Naglu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013792.2 | NP_038820.2 | 6 | 1845 | ||
| GenBank mRNA | AK009286.1 | - | 1 | 415 | ||
| AK143546.1 | - | 6 | 1847 | |||
| AK158774.1 | - | 4 | 2469 | |||
| AK164125.1 | - | 6 | 1824 | |||
| AK170758.1 | - | 6 | 1828 | |||
| AK178593.1 | - | 1 | 78 | |||
| AK208064.1 | - | 1 | 117 | |||
| BC055733.1 | - | 6 | 1819 | 6411530 | ||
| U85247.1 | - | 6 | 1814 | |||

Naglu
Genomic Map