siRNA Details
| Gene Symbol: Nagpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013796.3 | NP_038824.2 | 4, 5 | 821 | ||
| GenBank mRNA | AK032158.1 | - | 4, 5 | 822 | ||
| AK138035.1 | - | 4, 5 | 814 | |||
| AK155450.1 | - | 4, 5 | 800 | |||
| AK159629.1 | - | 4, 5 | 800 | |||
| AK162633.1 | - | 4, 5 | 803 | |||
| AK168830.1 | - | 3, 4 | 906 | |||
| AK170219.1 | - | 4, 5 | 803 | |||
| BC019640.1 | - | 3, 4 | 883 | |||
| BC039790.1 | - | 4, 5 | 813 | 4951805 | ||

Nagpa
Genomic Map