siRNA Details
| Gene Symbol: Smtnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177776.3 | NP_808444.1 | 8 | 1431 | ||
| GenBank mRNA | AK051585.1 | - | 8 | 1431 | ||
| AK085988.1 | - | 8 | 1306 | |||
| BC037993.1 | - | 8 | 1429 | 5064159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smtnl2| Gene Symbol: Smtnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177776.3 | NP_808444.1 | 8 | 1431 | ||
| GenBank mRNA | AK051585.1 | - | 8 | 1431 | ||
| AK085988.1 | - | 8 | 1306 | |||
| BC037993.1 | - | 8 | 1429 | 5064159 | ||