siRNA Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026585.3 | NP_080861.2 | 22, 23 | 2388 | ||
| GenBank mRNA | AK038318.1 | - | 22, 23 | 2366 | ||
| AK043764.1 | - | 11, 12 | 1233 | |||
| AK122320.1 | - | 21, 22 | 2394 | |||
| AK145716.1 | - | 22, 23 | 2389 | |||
| AK161181.1 | - | 22, 23 | 2338 | |||
| AK163765.1 | - | 21, 22 | 2376 | |||
| AK168388.1 | - | 22, 23 | 2377 | |||
| BC056942.1 | - | 22, 23 | 2315 | 6827177 | ||

Fam21
Genomic Map