siRNA Details
| Gene Symbol: Larp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138593.2 | NP_613059.2 | 12 | 1753 | ||
| GenBank mRNA | AK012726.1 | - | 1 | 55 | ||
| BC029178.1 | - | 4 | 426 | |||
| BC132266.1 | - | 4 | 424 | |||
| BC132268.1 | - | 4 | 424 | |||
| BM946683.1 | - | 6 | 621 | |||
| BU701061.1 | - | 2 | 379 | |||

Larp7
Genomic Map