siRNA Details
| Gene Symbol: Bag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013863.5 | NP_038891.4 | 4 | 1749 | ||
| GenBank mRNA | AB041583.1 | - | 4 | 1716 | ||
| AF130471.1 | - | 4 | 1482 | |||
| AK007414.1 | - | 4 | 1560 | |||
| AK016510.1 | - | 4 | 1556 | |||
| AK036675.1 | - | 4 | 1558 | |||
| AK171734.1 | - | 4 | 1691 | |||
| BC145765.1 | - | 4 | 1662 | 40131068 | ||

Bag3
Genomic Map