siRNA Details
Gene Symbol: Gm11946 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020625.1 | - | 1 | 71 |
Gene Symbol: Smtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159284.1 | NP_001152756.1 | 11 | 1284 | ||
NM_001284427.1 | NP_001271356.1 | 10 | 1233 | |||
NM_001284428.1 | NP_001271357.1 | 11 | 1341 | |||
NM_001284429.1 | NP_001271358.1 | 11 | 1286 | |||
NM_013870.3 | NP_038898.2 | 10 | 1233 | |||
GenBank mRNA | AF064236.1 | - | 10 | 1180 | ||
AJ010305.1 | - | 10 | 1180 | |||
AK030932.1 | - | 10 | 1234 | |||
AK085412.1 | - | 11 | 1248 | |||
AK138898.1 | - | 11 | 1298 | |||
AK166385.1 | - | 11 | 1301 | |||
AK170516.1 | - | 10 | 1193 | |||
BC010599.1 | - | 10 | 1174 | 3709078 | ||
BC066192.1 | - | 11 | 1162 | 30433616 | ||
BC069836.1 | - | 11 | 1162 | 30456568 | ||
BC069839.1 | - | 11 | 1162 | 30431675 |
