siRNA Details
| Gene Symbol: Fbxw2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164768.1 | NP_001158240.1 | 5 | 1092 | ||
| NM_001164769.1 | NP_001158241.1 | 7 | 1151 | |||
| NM_001164770.1 | NP_001158242.1 | 7 | 1059 | |||
| NM_013890.4 | NP_038918.3 | 7 | 1151 | |||
| GenBank mRNA | AF140683.1 | - | 5 | 1096 | ||
| AK033129.1 | - | 7 | 1105 | |||
| AK040764.1 | - | 7 | 1118 | |||
| AK078815.1 | - | 7 | 1059 | |||
| CB203503.1 | - | 2 | 87 | |||
| X54352.1 | - | 6 | 1081 | |||

Fbxw2
Genomic Map