siRNA Details
| Gene Symbol: Snai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013914.2 | NP_038942.1 | 1, 2 | 148 | ||
| GenBank mRNA | AF133714.1 | - | 1, 2 | 70 | ||
| AF195655.1 | - | 1, 2 | 70 | |||
| AK042234.1 | - | 1, 2 | 148 | |||
| AK089285.1 | - | 1, 2 | 137 | |||
| AK153633.1 | - | 1, 2 | 190 | |||
| BC100724.1 | - | 1, 2 | 97 | 40045286 | ||
| BC100726.1 | - | 1, 2 | 97 | 40045284 | ||
| BC100727.1 | - | 1, 2 | 97 | 40045283 | ||

Snai3
Genomic Map