siRNA Details
| Gene Symbol: Rnf19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013923.2 | NP_038951.1 | 2 | 881 | ||
| GenBank mRNA | AF120206.1 | - | 774 | |||
| AF120207.1 | - | 1 | 774 | |||
| AF360999.1 | - | 1 | 307 | |||
| AK082665.1 | - | 2 | 833 | |||
| AK147768.1 | - | 2 | 869 | |||
| AK166197.1 | - | 2 | 869 | |||
| BC040769.1 | - | 1 | 656 | 4457807 | ||
| CO432840.1 | - | 1 | 230 | |||

Rnf19a
Genomic Map