siRNA Details
| Gene Symbol: Cbx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013926.1 | NP_038954.1 | 5 | 824 | ||
| GenBank mRNA | AB221673.1 | - | 5 | 761 | ||
| AF180370.1 | - | 5 | 824 | |||
| AK051905.1 | - | 254 | ||||
| BC014815.1 | - | 5 | 774 | 4456898 | ||

Cbx8
Genomic Map