siRNA Details
| Gene Symbol: Pld5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195816.1 | NP_001182745.1 | 3 | 492 | ||
| NM_176916.4 | NP_795890.2 | 3 | 796 | |||
| GenBank mRNA | AK046294.1 | - | 3 | 759 | ||
| AK082725.1 | - | 3 | 393 | |||
| AK144116.1 | - | 3 | 785 | |||
| BC100428.1 | - | 3 | 235 | 30247182 | ||
| BU609501.1 | - | 3 | 461 | |||
| CK626039.1 | - | 3 | 492 | |||

Pld5
Genomic Map