siRNA Details
Gene Symbol: 6030419C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176921.3 | NP_795895.1 | 2, 3 | 1371 | ||
GenBank mRNA | AK031382.1 | - | 2, 3 | 1373 | ||
BC096614.1 | - | 1, 2 | 795 | 6468423 |
