siRNA Details
| Gene Symbol: 6030419C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176921.3 | NP_795895.1 | 3 | 1506 | ||
| GenBank mRNA | AK031382.1 | - | 3 | 1508 | ||
| BC096614.1 | - | 2 | 930 | 6468423 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6030419C18Rik| Gene Symbol: 6030419C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176921.3 | NP_795895.1 | 3 | 1506 | ||
| GenBank mRNA | AK031382.1 | - | 3 | 1508 | ||
| BC096614.1 | - | 2 | 930 | 6468423 | ||