siRNA Details
| Gene Symbol: Zbtb39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198035.1 | NP_932152.1 | 2 | 1726 | ||
| GenBank mRNA | AK135409.1 | - | 2 | 1719 | ||
| AK155700.1 | - | 2 | 1703 | |||
| AK172936.1 | - | 1 | 1479 | |||
| BC058661.1 | - | 2 | 1726 | 6409441 | ||
| BC060725.1 | - | 1 | 1335 | |||
| BC079569.1 | - | 2 | 1622 | 30536951 | ||

Zbtb39
Genomic Map