siRNA Details
| Gene Symbol: Fbxw21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177069.3 | NP_796043.1 | 4 | 413 | ||
| GenBank mRNA | AK087710.1 | - | 4 | 414 | ||
| AK087902.1 | - | 4 | 412 | |||
| AK135949.1 | - | 4 | 414 | |||
| AK136136.1 | - | 4 | 410 | |||
| BC099397.1 | - | 4 | 395 | 30936381 | ||

Fbxw21
Genomic Map