siRNA Details
| Gene Symbol: Fndc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177075.4 | NP_796049.1 | 2 | 82 | ||
| GenBank mRNA | AK081240.1 | - | 2 | 83 | ||
| BC104394.2 | - | 1 | 56 | 40050426 | ||
| BC104395.1 | - | 1 | 56 | 40050429 | ||
| BY728197.1 | - | 2 | 83 | |||
| Gene Symbol: Cyfip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037974.1 | - | 1 | 1103 | ||
| BC056974.1 | - | 24 | 3637 | 6831770 | ||

Fndc9
Genomic Map