siRNA Details
| Gene Symbol: Ptpn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177081.3 | NP_796055.1 | 8 | 847 | ||
| GenBank mRNA | AK041415.1 | - | 5 | 2888 | ||
| AK049175.1 | - | 7 | 698 | |||
| AK083305.1 | - | 8 | 847 | |||
| BC098475.1 | - | 8 | 901 | 30044369 | ||
| BC138194.1 | - | 7 | 783 | 8861213 | ||
| BC145190.1 | - | 7 | 762 | 9053741 | ||

Ptpn7
Genomic Map