siRNA Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045001.1 | - | 1 | 1111 | ||
| AK080905.1 | - | 1 | 938 | |||
| AK086551.1 | - | 1 | 889 | |||
| AK087678.1 | - | 2 | 973 | |||
| BC150968.1 | - | 1 | 847 | 9087880 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pten| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045001.1 | - | 1 | 1111 | ||
| AK080905.1 | - | 1 | 938 | |||
| AK086551.1 | - | 1 | 889 | |||
| AK087678.1 | - | 2 | 973 | |||
| BC150968.1 | - | 1 | 847 | 9087880 | ||