siRNA Details
| Gene Symbol: D930030D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086454.1 | - | 533 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D930030D11Rik| Gene Symbol: D930030D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086454.1 | - | 533 | |||