siRNA Details
| Gene Symbol: Tnfsf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177371.3 | NP_796345.3 | 3, 4 | 381 | ||
| GenBank mRNA | AF520786.1 | - | 3, 4 | 298 | ||
| AY764130.1 | - | 3, 4 | 381 | |||
| BC096494.1 | - | 3, 4 | 367 | 30740802 | ||
| BC138207.1 | - | 3, 4 | 346 | 8861226 | ||
| BC138208.1 | - | 3, 4 | 346 | 8861227 | ||

Tnfsf15
Genomic Map