siRNA Details
| Gene Symbol: Edaradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133643.4 | NP_598398.3 | 6 | 6218 | ||
| GenBank mRNA | AK041884.1 | - | 1 | 460 | ||
| AK162653.1 | - | 1 | 3786 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Edaradd| Gene Symbol: Edaradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133643.4 | NP_598398.3 | 6 | 6218 | ||
| GenBank mRNA | AK041884.1 | - | 1 | 460 | ||
| AK162653.1 | - | 1 | 3786 | |||