siRNA Details
Gene Symbol: Edaradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133643.4 | NP_598398.3 | 6 | 6378 | ||
GenBank mRNA | AK041884.1 | - | 1 | 620 | ||
AK162653.1 | - | 1 | 3946 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Edaradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133643.4 | NP_598398.3 | 6 | 6378 | ||
GenBank mRNA | AK041884.1 | - | 1 | 620 | ||
AK162653.1 | - | 1 | 3946 |