siRNA Details
| Gene Symbol: Pkd2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181422.3 | NP_852087.2 | 3 | 602 | ||
| GenBank mRNA | AB290927.1 | - | 3 | 434 | ||
| AF271381.1 | - | 3 | 568 | |||
| AK046772.1 | - | 3 | 603 | |||
| BC116297.2 | - | 3 | 533 | 40094997 | ||
| BC116323.2 | - | 3 | 533 | 40096335 | ||

Pkd2l1
Genomic Map