siRNA Details
| Gene Symbol: Tbc1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198664.3 | NP_941066.3 | 12, 13 | 2736 | ||
| GenBank mRNA | AK148289.1 | - | 12, 13 | 2652 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d2| Gene Symbol: Tbc1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198664.3 | NP_941066.3 | 12, 13 | 2736 | ||
| GenBank mRNA | AK148289.1 | - | 12, 13 | 2652 | ||