siRNA Details
| Gene Symbol: Fhad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177868.4 | NP_808536.2 | 23 | 3138 | ||
| GenBank mRNA | AK148490.1 | - | 8 | 1223 | ||
| BC038692.1 | - | 5 | 721 | 1496088 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fhad1| Gene Symbol: Fhad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177868.4 | NP_808536.2 | 23 | 3138 | ||
| GenBank mRNA | AK148490.1 | - | 8 | 1223 | ||
| BC038692.1 | - | 5 | 721 | 1496088 | ||