siRNA Details
| Gene Symbol: Mblac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177878.3 | NP_808546.1 | 1 | 447 | ||
| GenBank mRNA | AK049381.1 | - | 1 | 448 | ||
| BC038925.1 | - | 1 | 428 | 5374241 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mblac1| Gene Symbol: Mblac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177878.3 | NP_808546.1 | 1 | 447 | ||
| GenBank mRNA | AK049381.1 | - | 1 | 448 | ||
| BC038925.1 | - | 1 | 428 | 5374241 | ||