siRNA Details
| Gene Symbol: Tcrb-J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017898.1 | - | 4 | 351 | ||
| M11456.1 | - | 2 | 172 | |||
| M13676.1 | - | 2 | 145 | |||
| M16679.1 | - | 2 | 145 | |||
| M20877.1 | - | 1 | 94 | |||
| M20878.1 | - | 1 | 94 | |||
| U46842.1 | - | 2 | 145 | |||
| X00438.1 | - | 2 | 167 | |||
| X02778.1 | - | 2 | 121 | |||
| X02779.1 | - | 2 | 145 | |||

Tcrb-J
Genomic Map