siRNA Details
| Gene Symbol: Gcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177884.3 | NP_808552.2 | 15 | 2036 | ||
| GenBank mRNA | AK038369.1 | - | 16 | 2023 | ||
| AK050789.1 | - | 15 | 1978 | |||
| AK078171.1 | - | 15 | 2026 | |||
| AK136387.1 | - | 13 | 1638 | |||

Gcfc2
Genomic Map