siRNA Details
| Gene Symbol: Lcn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178036.4 | NP_828875.1 | 1 | 73 | ||
| GenBank mRNA | AF435737.1 | - | 1 | 73 | ||
| BC119103.1 | - | 1 | 41 | 8733852 | ||
| BC120657.1 | - | 1 | 41 | 40129580 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lcn10| Gene Symbol: Lcn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178036.4 | NP_828875.1 | 1 | 73 | ||
| GenBank mRNA | AF435737.1 | - | 1 | 73 | ||
| BC119103.1 | - | 1 | 41 | 8733852 | ||
| BC120657.1 | - | 1 | 41 | 40129580 | ||