siRNA Details
| Gene Symbol: Tfap2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198960.2 | NP_945198.2 | 2, 3 | 783 | ||
| GenBank mRNA | AY325902.1 | - | 2, 3 | 623 | ||
| BI134735.1 | - | 1, 2 | 193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tfap2e| Gene Symbol: Tfap2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198960.2 | NP_945198.2 | 2, 3 | 783 | ||
| GenBank mRNA | AY325902.1 | - | 2, 3 | 623 | ||
| BI134735.1 | - | 1, 2 | 193 | |||